AMBH580648DB Ambeed, Inc.

AS-602801

  • MDL number MFCD08272350
  • Cas Number : 848344-36-5
  • Molecular Weight : 457.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/848344-36-5.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/848344-36-5.html
pictograms GHS07
hstatements H302-H315-H332
Availability details for
Quantity
  • Availability
  • Add to Cart