AMBH580648DD Ambeed, Inc.

tert-Butyl 3,5-dimethyl-4-oxopiperidine-1-carboxylate

  • MDL number MFCD14635714
  • Cas Number : 475085-34-8
  • Molecular Weight : 227.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/475085-34-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/475085-34-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart