AMBH580648DE Ambeed, Inc.

(6R,7R)-7-Amino-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

  • MDL number MFCD06656104
  • Cas Number : 30246-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/30246-33-4.html

Safety Information

MSDS https://www.ambeed.com/products/30246-33-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart