AMBH580648E1 Ambeed, Inc.

3-Methoxy-2-methylbenzaldehyde

  • MDL number MFCD11044581
  • Cas Number : 56724-03-9
  • Molecular Weight : 150.1700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form liquid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/56724-03-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501
MSDS https://www.ambeed.com/products/56724-03-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart