AMBH580648E3 Ambeed, Inc.

(2S,4R)-Methyl 4-(tert-butoxycarbonylamino)pyrrolidine-2-carboxylate

  • MDL number MFCD08704530
  • Cas Number : 473806-21-2
  • Molecular Weight : 244.2875

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/473806-21-2.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/473806-21-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart