AMBH580648E8 Ambeed, Inc.

3-Methoxy-1-N,1-N-dimethylbenzene-1,4-diamine

  • MDL number MFCD16249531
  • Cas Number : 7474-82-0
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 266.5�t 760 mmHg
Iupac Name 2-methoxy-N~4~,N~4~-dimethyl-1,4-benzenediamine
Inchi Code 1S/C9H14N2O/c1-11(2)7-4-5-8(10)9(6-7)12-3/h4-6H,10H2,1-3H3
Inchi Key PXABZNHUEJWLMA-UHFFFAOYSA-N

Description

Synonym 3-Methoxy-N1,N1-dimethylbenzene-1,4-diamine
Purity 97%
Flash Point 115
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 266.5�t 760 mmHg
Product Link https://www.ambeed.com/products/7474-82-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7474-82-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart