AMBH580648EB Ambeed, Inc.

PREDNISONEACETATE

  • MDL number MFCD00200229
  • Cas Number : 125-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/125-10-0.html

Safety Information

MSDS https://www.ambeed.com/products/125-10-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart