AMBH580648F0 Ambeed, Inc.

Diphenyl ((6-methylpyridin-2-yl)(phenylamino)methyl)phosphonate

  • MDL number MFCD18632870
  • Cas Number : 614750-85-5
  • Molecular Weight : 430.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/614750-85-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/614750-85-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart