AMBH580648F6 Ambeed, Inc.

(2S)-2-([(9H-FLUOREN-9-YLMETHOXY)CARBONYL]AMINO)-3-(QUINOLIN-3-YL)PROPANOIC ACID

  • MDL number MFCD01632017
  • Cas Number : 281655-61-6
  • Molecular Weight : 438.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/281655-61-6.html

Safety Information

MSDS https://www.ambeed.com/products/281655-61-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart