AMBH580648FC Ambeed, Inc.

Sucrose octaacetate

  • MDL number MFCD00006623
  • Cas Number : 126-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/126-14-7.html

Safety Information

MSDS https://www.ambeed.com/products/126-14-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart