AMBH58064908 Ambeed, Inc.

Cbz-L-Isoleucine

  • MDL number MFCD00027064
  • Cas Number : 3160-59-6
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3160-59-6.html

Safety Information

MSDS https://www.ambeed.com/products/3160-59-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart