AMBH5806490B Ambeed, Inc.

1-(2-Fluorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09950609
  • Cas Number : 1134310-63-6
  • Molecular Weight : 206.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1134310-63-6.html

Safety Information

MSDS https://www.ambeed.com/products/1134310-63-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart