AMBH5806490F Ambeed, Inc.

(S)-1-(4-(4Methylthiazol-5-yl)phenyl)ethan-1-amine HCl

  • MDL number MFCD30741910
  • Cas Number : 1948273-01-5
  • Molecular Weight : 254.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1948273-01-5.html

Safety Information

MSDS https://www.ambeed.com/products/1948273-01-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart