AMBH58064911 Ambeed, Inc.

7-Iodoisobenzofuran-1(3H)-one

  • MDL number MFCD18204743
  • Cas Number : 105694-46-0
  • Molecular Weight : 260.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-iodo-2-benzofuran-1(3H)-one
Inchi Code 1S/C8H5IO2/c9-6-3-1-2-5-4-11-8(10)7(5)6/h1-3H,4H2
Inchi Key HCQIZIWFLKWDFR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/105694-46-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/105694-46-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart