AMBH58064912 Ambeed, Inc.

N,N-Diethyl-3-methylaniline

  • MDL number MFCD00035795
  • Cas Number : 91-67-8
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/91-67-8.html

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312+P330-P302+P352+P312+P362+P364-P304+P340+P312-P305+P351+P338+P337+P313-P501
MSDS https://www.ambeed.com/products/91-67-8.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart