AMBH58064914 Ambeed, Inc.

tert-Butyl 4-(methylcarbamoyl)piperidine-1-carboxylate

  • MDL number MFCD06411659
  • Cas Number : 544696-01-7
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/544696-01-7.html

Safety Information

MSDS https://www.ambeed.com/products/544696-01-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart