AMBH58064916 Ambeed, Inc.

(S)-Di-Tert-Butyl 2-(3-((S)-6-Amino-1-Tert-Butoxy-1-Oxohexan-2-Yl)ureido)pentanedioate

  • MDL number MFCD28898592
  • Cas Number : 1025796-31-9
  • Molecular Weight : 487.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1025796-31-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1025796-31-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart