AMBH5806491B Ambeed, Inc.

Sodium 2-cyanoacetate

  • MDL number MFCD00067028
  • Cas Number : 1071-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1071-36-9.html

Safety Information

MSDS https://www.ambeed.com/products/1071-36-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart