AMBH58064923 Ambeed, Inc.

2,2'-(m-Tolylazanediyl)diethanol

  • MDL number MFCD00020578
  • Cas Number : 91-99-6
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/91-99-6.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P312
MSDS https://www.ambeed.com/products/91-99-6.html
pictograms GHS06
hstatements H301-H302-H311-H312-H317-H332
Availability details for
Quantity
  • Availability
  • Add to Cart