AMBH58064925 Ambeed, Inc.

2-Phenoxyethyl acetate

  • MDL number MFCD00059336
  • Cas Number : 6192-44-5
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 95.3
Storage temp. Sealed in dry,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 243.6°C at 760 mmHg
Product Link https://www.ambeed.com/products/6192-44-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/6192-44-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart