AMBH58064926 Ambeed, Inc.

5,6,7,8-Tetrahydroimidazo[1,2-a]pyridin-7-ol

  • MDL number MFCD28654766
  • Cas Number : 1785629-71-1
  • Molecular Weight : 138.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1785629-71-1.html

Safety Information

MSDS https://www.ambeed.com/products/1785629-71-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart