AMBH58064932 Ambeed, Inc.

5-Fluoro-1-(2-fluorobenzyl)-3-iodo-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD28502338
  • Cas Number : 1350653-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1350653-24-5.html

Safety Information

MSDS https://www.ambeed.com/products/1350653-24-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart