AMBH58064937 Ambeed, Inc.

N-BUTYLTINHYDROXIDEOXIDE

  • MDL number MFCD00013927
  • Cas Number : 2273-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/2273-43-0.html

Safety Information

MSDS https://www.ambeed.com/products/2273-43-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart