AMBH5806493C Ambeed, Inc.

3-Hydroxy-2-naphthonitrile

  • MDL number MFCD17012306
  • Cas Number : 52449-77-1
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/52449-77-1.html

Safety Information

MSDS https://www.ambeed.com/products/52449-77-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart