AMBH5806493F Ambeed, Inc.

Methyl 3-amino-1-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD16619854
  • Cas Number : 64182-19-0
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/64182-19-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/64182-19-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart