AMBH58064949 Ambeed, Inc.

(R)-2,2-Dimethyl-1,3-dioxolane-4-carbaldehyde

  • MDL number MFCD00269682
  • Cas Number : 15186-48-8
  • Molecular Weight : 130.1418

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 50
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/15186-48-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/15186-48-8.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart