AMBH58064950 Ambeed, Inc.

(R)-3-((tert-Butoxycarbonyl)amino)-4,4-diphenylbutanoic acid

  • MDL number MFCD02094565
  • Cas Number : 332062-06-3
  • Molecular Weight : 355.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/332062-06-3.html

Safety Information

MSDS https://www.ambeed.com/products/332062-06-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart