AMBH58064951 Ambeed, Inc.

Bis(pyridin-4-ylmethyl)amine

  • MDL number MFCD00868302
  • Cas Number : 1539-39-5
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1539-39-5.html

Safety Information

MSDS https://www.ambeed.com/products/1539-39-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart