AMBH58064952 Ambeed, Inc.

3-Isopropylbenzene-1-sulfonyl chloride

  • MDL number MFCD09730000
  • Cas Number : 71530-58-0
  • Molecular Weight : 218.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/71530-58-0.html

Safety Information

MSDS https://www.ambeed.com/products/71530-58-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart