AMBH5806495E Ambeed, Inc.

3-Pentanone p-Toluenesulfonylhydrazone

  • MDL number MFCD03367534
  • Cas Number : 28495-72-9
  • Molecular Weight : 254.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N'-(1-ethylpropylidene)-4-methylbenzenesulfonohydrazide
Inchi Code 1S/C12H18N2O2S/c1-4-11(5-2)13-14-17(15,16)12-8-6-10(3)7-9-12/h4,6-9,13-14H,5H2,1-3H3
Inchi Key JRRALSSVNFGLIP-UHFFFAOYSA-N

Description

Synonym 4-Methyl-N'-(pentan-3-ylidene)benzenesulfonohydrazide
Purity 98%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/28495-72-9.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/28495-72-9.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart