AMBH58064961 Ambeed, Inc.

4,5-Dichlorophthalic Anhydride

  • MDL number MFCD00075034
  • Cas Number : 942-06-3
  • Molecular Weight : 217.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-187°C
Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/942-06-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/942-06-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart