AMBH5806496A Ambeed, Inc.

(E)-4-(4-Methylpiperazin-1-yl)but-2-enoic acid

  • MDL number MFCD16712914
  • Cas Number : 1323199-65-0
  • Molecular Weight : 184.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1323199-65-0.html

Safety Information

MSDS https://www.ambeed.com/products/1323199-65-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart