AMBH58064970 Ambeed, Inc.

3-Iodobenzene-1,2-diamine

  • MDL number MFCD12028717
  • Cas Number : 34446-43-0
  • Molecular Weight : 234.0377

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/34446-43-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/34446-43-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart