AMBH58064972 Ambeed, Inc.

Benzo[4,5]thieno[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD01830098
  • Cas Number : 36822-08-9
  • Molecular Weight : 202.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/36822-08-9.html

Safety Information

MSDS https://www.ambeed.com/products/36822-08-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart