AMBH58064973 Ambeed, Inc.

[1,1'-Biphenyl]-2-ylmethanamine

  • MDL number MFCD00270123
  • Cas Number : 1924-77-2
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1924-77-2.html

Safety Information

MSDS https://www.ambeed.com/products/1924-77-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart