AMBH58064976 Ambeed, Inc.

MBX-2982

  • MDL number MFCD22628771
  • Cas Number : 1037792-44-1
  • Molecular Weight : 448.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1037792-44-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1037792-44-1.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart