AMBH58064983 Ambeed, Inc.

6-(Trifluoromethyl)-1H-benzo[d]imidazole

  • MDL number MFCD06659629
  • Cas Number : 326-55-6
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 98
Flash Point 159
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 339.3�t 760 mmHg
Product Link https://www.ambeed.com/products/326-55-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/326-55-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart