AMBH5806498B Ambeed, Inc.

3,4-Dihydro-2H-pyrano[3,2-b]pyridin-4-amine

  • MDL number MFCD13180519
  • Cas Number : 502612-49-9
  • Molecular Weight : 150.1778

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/502612-49-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/502612-49-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart