AMBH58064999 Ambeed, Inc.

2-Chloro-7-(trifluoromethyl)-1H-benzo[d]imidazole

  • MDL number MFCD22562508
  • Cas Number : 1075753-27-3
  • Molecular Weight : 220.5790

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1075753-27-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1075753-27-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart