AMBH580649A0 Ambeed, Inc.

3-Ethoxy-5-(trifluoromethyl)picolinic acid

  • MDL number MFCD00205857
  • Cas Number : 257862-54-7
  • Molecular Weight : 235.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-ethoxy-5-(trifluoromethyl)picolinic acid
Inchi Code 1S/C9H8F3NO3/c1-2-16-6-3-5(9(10,11)12)4-13-7(6)8(14)15/h3-4H,2H2,1H3,(H,14,15)
Inchi Key OZTWGNULSXOKAM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/257862-54-7.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/257862-54-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart