AMBH580649A3 Ambeed, Inc.

Ethyl 2-amino-3,4,5-trifluorobenzoate

  • MDL number MFCD12159863
  • Cas Number : 1184500-46-6
  • Molecular Weight : 219.1605

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form Liquid or solid
Shipping Temperature Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1184500-46-6.html

Safety Information

signalword Danger
pcodes P261-P264-P270-P271-P280-P302+P352-P304+P340-P310-P330-P361-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/1184500-46-6.html
pictograms GHS06
hstatements H301-H311-H331
Availability details for
Quantity
  • Availability
  • Add to Cart