AMBH580649A8 Ambeed, Inc.

4-Cyclopropyl-2-fluoro-6-methylbenzamide

  • MDL number MFCD20726776
  • Cas Number : 1242156-52-0
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1242156-52-0.html

Safety Information

MSDS https://www.ambeed.com/products/1242156-52-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart