AMBH580649B4 Ambeed, Inc.

2-Chloro-4-methoxy-5-nitropyrimidine

  • MDL number MFCD00955634
  • Cas Number : 282102-07-2
  • Molecular Weight : 189.5566

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/282102-07-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/282102-07-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart