AMBH580649BF Ambeed, Inc.

1-(3-Bromoprop-2-ynyl)piperidine

  • MDL number MFCD31567073
  • Cas Number : 19977-51-6
  • Molecular Weight : 202.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromoprop-2-yn-1-yl)piperidine
Inchi Code 1S/C8H12BrN/c9-5-4-8-10-6-2-1-3-7-10/h1-3,6-8H2
Inchi Key IPXFLXHWSHBHKU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/19977-51-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/19977-51-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart