AMBH580649C1 Ambeed, Inc.

1-Phenyl-1-hexyne

  • MDL number MFCD00039969
  • Cas Number : 1129-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1129-65-3.html

Safety Information

MSDS https://www.ambeed.com/products/1129-65-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart