AMBH580649C7 Ambeed, Inc.

(S)-4-(tert-Butyl)-2-(2-(diphenylphosphino)phenyl)-4,5-dihydrooxazole

  • MDL number MFCD09998277
  • Cas Number : 148461-16-9
  • Molecular Weight : 387.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/148461-16-9.html

Safety Information

MSDS https://www.ambeed.com/products/148461-16-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart