AMBH580649CA Ambeed, Inc.

Methyl 2-bromo-5-fluoro-4-methylbenzoate

  • MDL number MFCD28739724
  • Cas Number : 1204304-98-2
  • Molecular Weight : 247.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1204304-98-2.html

Safety Information

MSDS https://www.ambeed.com/products/1204304-98-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart