AMBH580649CB Ambeed, Inc.

(S)-2-Amino-4-(4-hydroxyphenyl)butanoic acid

  • MDL number MFCD03490505
  • Cas Number : 221243-01-2
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/221243-01-2.html

Safety Information

MSDS https://www.ambeed.com/products/221243-01-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart