AMBH580649CC Ambeed, Inc.

1-(3-Chloropyridin-4-yl)ethanone

  • MDL number MFCD10000759
  • Cas Number : 78790-82-6
  • Molecular Weight : 155.5816

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/78790-82-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/78790-82-6.html
pictograms GHS07
hstatements H302-H315-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart