AMBH580649D4 Ambeed, Inc.

N-(2,4-Dimethylphenyl)-3-oxobutyramide

  • MDL number MFCD00039836
  • Cas Number : 97-36-9
  • Molecular Weight : 205.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/97-36-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/97-36-9.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart